3-Amino-N,N-dimethylbenzylamine 96%

Supplier: Apollo Scientific

Synonyms: 3-((Dimethylamino)methyl)aniline

OR919226-1G
APOSOR919226-1GEA 119 EUR
APOSOR919226-1G
3-Amino-N,N-dimethylbenzylamine 96%
3-Amino-N,N-dimethylbenzylamine
Formula: C₉H₁₄N₂
Storage Temperature: Ambient
MDL Number: MFCD00036095
CAS Number: 27958-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR