1-(2-Chloroethoxy)propane 98%

Supplier: Apollo Scientific

Synonyms: 2-Chloroethyln-propylether, 2-Chloroethyl Propyl ether, 2-Propoxyethyl chloride

OR919962-25G OR919962-100G
APOSOR919962-25GEA 72.5 EUR
APOSOR919962-25G APOSOR919962-100G
1-(2-Chloroethoxy)propane 98%
1-(2-Chloroethoxy)propane
Formula: ClCH₂CH₂OCH₂CH₂CH₃
Storage Temperature: Ambient
MDL Number: MFCD00059052
CAS Number: 42149-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR