Bis(2-chloroethyl)ether 95%

Supplier: Apollo Scientific

Synonyms: 2-Chloroethyl ether

OR922075-100G OR922075-1KG OR922075-500G
APOSOR922075-100GEA 109 EUR
APOSOR922075-100G APOSOR922075-1KG APOSOR922075-500G
Bis(2-chloroethyl)ether 95%
Bis(2-chloroethyl) ether
Formula: C₄H₈Cl₂O
Boiling Pt: 178 °C/760mmHg
Melting Pt: –52 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00000975
CAS Number: 111-44-4
UN: 1916
ADR: 6.1+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR