4-Methylcyclohexylamine (cis- and trans- mixture)

Supplier: Apollo Scientific

Synonyms: 1-Amino-4-methylcyclohexane

OR924724-1G OR924724-25G OR924724-5G
APOSOR924724-1GEA 29 EUR
APOSOR924724-1G APOSOR924724-25G APOSOR924724-5G
4-Methylcyclohexylamine (cis- and trans- mixture)
4-Methylcyclohexylamine (cis- and trans- mixture)
Formula: C₇H₁₅N
Boiling Pt: 148…154 °C
Density: 0.855 g/cm³ (25 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00001495
CAS Number: 6321-23-9
UN: 2734
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR