Morpholin-3-ylmethanol 96%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Hydroxymethylmorpholine

OR926444-1G OR926444-5G
APOSOR926444-1GEA 94.8 EUR
APOSOR926444-1G APOSOR926444-5G
Morpholin-3-ylmethanol 96%
Morpholin-3-ylmethanol
Formula: C₅H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD04038650
CAS Number: 106910-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR