2,5-Dimethyltetrahydrofuran 95%

Supplier: Apollo Scientific

OR928860-25ML
APOSOR928860-25MLEA 248 EUR
APOSOR928860-25ML
2,5-Dimethyltetrahydrofuran 95%
2,5-Dimethyltetrahydrofuran
Formula: C₆H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00005369
CAS Number: 1003-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR