3-Dimethoxymethyl-2-methoxy-pyridine 95%

Supplier: Apollo Scientific

OR952352-1G OR952352-250MG
APOSOR952352-1GEA 451 EUR
APOSOR952352-1G APOSOR952352-250MG
3-Dimethoxymethyl-2-methoxy-pyridine 95%
3-Dimethoxymethyl-2-methoxypyridine
Formula: C₉H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD07366720
CAS Number: 869735-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR