4-Morpholin-4-ylmethylthiazol-2-ylamine 97%

Supplier: Apollo Scientific

Synonyms: 4-(Morpholinomethyl)thiazol-2-amine

OR965574-100MG OR965574-1G OR965574-250MG
APOSOR965574-100MGEA 69.6 EUR
APOSOR965574-100MG APOSOR965574-1G APOSOR965574-250MG
4-Morpholin-4-ylmethylthiazol-2-ylamine 97%
4-Morpholin-4-ylmethylthiazol-2-ylamine
Formula: C₈H₁₃N₃OS
Storage Temperature: Ambient
MDL Number: MFCD06340096
CAS Number: 3008-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR