5-Chloro-2,3-difluoropyridine 97%

Supplier: Apollo Scientific
Warning

PC10516-100G PC10516-25G PC10516-5G
APOSPC10516-100GEA 30.6 EUR
APOSPC10516-100G APOSPC10516-25G APOSPC10516-5G
5-Chloro-2,3-difluoropyridine 97%
5-Chloro-2,3-difluoropyridine
Formula: C5H2ClF2N
Boiling Pt: 135 °C
Density: 1.442 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039314
CAS Number: 89402-43-7
EINECS: 410-090-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR