5-Fluoro-7-(methylsulphonyl)-1H-indole-2-carboxylic acid 95+%

Supplier: Apollo Scientific
Warning

PC11193-1G PC11193-10G
APOSPC11193-1GEA 705 EUR
APOSPC11193-1G APOSPC11193-10G
5-Fluoro-7-(methylsulphonyl)-1H-indole-2-carboxylic acid 95+%
5-Fluoro-7-(methylsulphonyl)-1H-indole-2-carboxylic acid
Formula: C₁₀H₈FNO₄S
Melting Pt: 270…272 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04037958
CAS Number: 849035-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR