3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 95%

Supplier: Apollo Scientific
Warning

PC1931E-5G
APOSPC1931E-5GEA 29 EUR
APOSPC1931E-5G
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 95%
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃NO
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00153095
CAS Number: 76041-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR