2-Chloro-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine

Supplier: Apollo Scientific

PC200288-1G PC200288-500MG
APOSPC200288-1GEA 471 EUR
APOSPC200288-1G APOSPC200288-500MG
2-Chloro-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine
2-Chloro-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine
Formula: C₈H₃ClF₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD14581666
CAS Number: 1203898-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR