2,3-Dichlorooctafluorobutane 97%

Supplier: Apollo Scientific
Warning

PC2490-1G
APOSPC2490-1GEA 21.9 EUR
APOSPC2490-1G
2,3-Dichlorooctafluorobutane 97%
2,3-Dichlorooctafluorobutane
Formula: C₄Cl₂F₈
Boiling Pt: 63 °C
Melting Pt: –68 °C
Density: 1.6801 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00018064
CAS Number: 355-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR