2,3-Difluoroaniline 98%

Supplier: Apollo Scientific
Warning

PC2588-100G PC2588-25G PC2588-5G
APOSPC2588-100GEA 302 EUR
APOSPC2588-100G APOSPC2588-25G APOSPC2588-5G
2,3-Difluoroaniline 98%
2,3-Difluoroaniline
Formula: C₆H₅F₂N
Boiling Pt: 68…69 °C/12mmHg
Density: 1.274 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00010298
CAS Number: 4519-40-8
EINECS: 224-847-2
UN: 2941
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR