4'-Methoxy-3'-(trifluoromethyl)acetophenone 98+%

Supplier: Apollo Scientific
Warning

Synonyms: 1-(4-Methoxy-3-(trifluoromethyl)phenyl)ethanone

PC2631-1G PC2631-25G PC2631-5G
APOSPC2631-1GEA 21.9 EUR
APOSPC2631-1G APOSPC2631-25G APOSPC2631-5G
4'-Methoxy-3'-(trifluoromethyl)acetophenone 98+%
4'-Methoxy-3'-(trifluoromethyl)acetophenone
Formula: C₁₀H₉F₃O₂
Melting Pt: 56…61 °C
Storage Temperature: Ambient
MDL Number: MFCD01091010
CAS Number: 149105-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR