4-Bromo-2,5-difluoroiodobenzene

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-2,5-difluoro-4-iodobenzene

PC2687-5G PC2687-1G PC2687-25G
APOSPC2687-5GEA 37.7 EUR
APOSPC2687-5G APOSPC2687-1G APOSPC2687-25G
4-Bromo-2,5-difluoroiodobenzene
4-Bromo-2,5-difluoroiodobenzene
Formula: C₆H₂BrF₂I
Melting Pt: 54…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04973906
CAS Number: 145349-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR