6-Bromo-2-fluoropyridin-3-amine ≥97%

Supplier: Apollo Scientific
Warning

PC28224-250MG
APOSPC28224-250MGEA 55.1 EUR
APOSPC28224-250MG
6-Bromo-2-fluoropyridin-3-amine ≥97%
6-Bromo-2-fluoropyridin-3-amine
Formula: C₅H₄BrFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD13659314
CAS Number: 850220-97-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR