(3,5-Difluorophenyl)acetonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3,5-Difluorobenzyl cyanide, 2-(3,5-Difluorophenyl)acetonitrile

PC2874EL-100G PC2874EL-10G PC2874EL-25G
APOSPC2874EL-100GEA 184 EUR
APOSPC2874EL-100G APOSPC2874EL-10G APOSPC2874EL-25G
(3,5-Difluorophenyl)acetonitrile 98%
(3,5-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
Melting Pt: 34…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00061278
CAS Number: 122376-76-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR