4'-Fluoroacetophenone 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone

PC2887-100G PC2887-1KG PC2887-250G PC2887-500G
APOSPC2887-100GEA 29 EUR
APOSPC2887-100G APOSPC2887-1KG APOSPC2887-250G APOSPC2887-500G
4'-Fluoroacetophenone 95%
4'-Fluoroacetophenone
Formula: C₈H₇FO
Boiling Pt: 194…196 °C
Melting Pt: 4 °C
Density: 1.138 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00000354
CAS Number: 403-42-9
EINECS: 206-960-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR