2-Chloro-N-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]acetamide

Supplier: Apollo Scientific

PC300674-1G
APOSPC300674-1GEA 359 EUR
APOSPC300674-1G
2-Chloro-N-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]acetamide
2-Chloro-N-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]acetamide
Formula: C₉H₈ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD13194594
CAS Number: 1308649-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR