Methyl 2,5-difluoro-4-piperidin-1-ylbenzoate

Supplier: Apollo Scientific

PC300791-1G PC300791-250MG PC300791-5G
APOSPC300791-1GEA 290 EUR
APOSPC300791-1G APOSPC300791-250MG APOSPC300791-5G
Methyl 2,5-difluoro-4-piperidin-1-ylbenzoate
Methyl 2,5-difluoro-4-piperidin-1-ylbenzoate
Formula: C₁₃H₁₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD28103352
CAS Number: 1858241-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR