4-[8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile

Supplier: Apollo Scientific

PC300865-1G
APOSPC300865-1GEA 342 EUR
APOSPC300865-1G
4-[8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile
4-[8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile
Formula: C₁₅H₇ClF₃N₃
MW: 321.684 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28992260
CAS Number: 1980053-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR