2,3-Difluoro-4-methoxyphenylacetonitrile 97%

Supplier: Apollo Scientific

Synonyms: 2-(2,3-Difluoro-4-methoxyphenyl)acetonitrile

PC302153-1G PC302153-5G
APOSPC302153-1GEA 215 EUR
APOSPC302153-1G APOSPC302153-5G
2,3-Difluoro-4-methoxyphenylacetonitrile 97%
2,3-Difluoro-4-methoxyphenylacetonitrile
Formula: C₉H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD06660320
CAS Number: 886503-74-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR