3-Methoxy-4-(trifluoromethyl)benzyl alcohol 98%

Supplier: Apollo Scientific

PC303070-10G PC303070-5G PC303070-25G
APOSPC303070-10GEA 459 EUR
APOSPC303070-10G APOSPC303070-5G APOSPC303070-25G
3-Methoxy-4-(trifluoromethyl)benzyl alcohol 98%
3-Methoxy-4-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18398590
CAS Number: 276861-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR