3,4-Dichloro-5-(trifluoromethyl)acetophenone 97%

Supplier: Apollo Scientific
Warning

PC303166-1G
APOSPC303166-1GEA 300 EUR
APOSPC303166-1G
3,4-Dichloro-5-(trifluoromethyl)acetophenone 97%
3,4-Dichloro-5-(trifluoromethyl)acetophenone
Formula: C₉H₅Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD18394788
CAS Number: 1261645-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR