3-Bromo-4-methyl-5-(trifluoromethyl)benzyl alcohol

Supplier: Apollo Scientific

PC303600-1G PC303600-5G
APOSPC303600-1GEA 314 EUR
APOSPC303600-1G APOSPC303600-5G
3-Bromo-4-methyl-5-(trifluoromethyl)benzyl alcohol
3-Bromo-4-methyl-5-(trifluoromethyl)benzyl alcohol
Formula: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD32640173
CAS Number: 2385206-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR