2-Chloroethyl 2,2,2-trifluoroethyl ether 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(2-Chloroethoxy)-1,1,1-trifluoroethane, 5-Chloro-1,1,1-trifluoro-3-oxapentane

PC3546-1G PC3546-5G
APOSPC3546-1GEA 121 EUR
APOSPC3546-1G APOSPC3546-5G
2-Chloroethyl 2,2,2-trifluoroethyl ether 97%
2-Chloroethyl 2,2,2-trifluoroethyl ether
Formula: C₄H₆ClF₃O
Boiling Pt: 114…115 °C
Density: 1.303 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00462515
CAS Number: 1537-70-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR