5-Chloro-3-fluoro-2-(methylamino)pyridine

Supplier: Apollo Scientific
Warning

Synonyms: 5-Chloro-3-fluoro-N-methylpyridin-2-amine

PC3655-1G
APOSPC3655-1GEA 176 EUR
APOSPC3655-1G
5-Chloro-3-fluoro-2-(methylamino)pyridine
5-Chloro-3-fluoro-2-(methylamino)pyridine
Formula: C₆H₆ClFN₂
Storage Temperature: Ambient
MDL Number: MFCD09878437
CAS Number: 220714-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR