Ethyl-2-(4-(trifluoromethyl)phenyl)acetate 96%

Supplier: Apollo Scientific

Synonyms: Ethyl 2-(4-(trifluoromethyl)phenyl)acetate, Ethyl 4-(Trifluoromethyl)phenylacetate, 4-(Trifluoromethyl)phenylacetic acid ethyl ester

PC3679-25G
APOSPC3679-25GEA 150 EUR
APOSPC3679-25G
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate 96%
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate
Formula: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07779351
CAS Number: 721-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR