1-Fluoro-2-(2-nitrovinyl)benzene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoro-beta-nitrostyrene, 1-Fluoro-2-(2-nitroethenyl)benzene

PC3988-5G
APOSPC3988-5GEA 159 EUR
APOSPC3988-5G
1-Fluoro-2-(2-nitrovinyl)benzene 98%
1-Fluoro-2-(2-nitrovinyl)benzene
Formula: C₈H₆FNO₂
Melting Pt: 55…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042451
CAS Number: 399-25-7
EINECS: 206-915-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR