1-(3-Bromo-5-fluoropyridin-2-yl)-2,2,2-trifluoroethanone

Supplier: Apollo Scientific
Warning

PC400147-1G PC400147-250MG
APOSPC400147-1GEA 2030 EUR
APOSPC400147-1G APOSPC400147-250MG
1-(3-Bromo-5-fluoropyridin-2-yl)-2,2,2-trifluoroethanone
1-(3-Bromo-5-fluoropyridin-2-yl)-2,2,2-trifluoroethanone
Formula: C₇H₂BrF₄NO
Storage Temperature: Ambient
MDL Number: MFCD31621151
CAS Number: 2231675-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR