1-Nitro-3-(pentafluoroethyl)benzene

Supplier: Apollo Scientific

PC408831-1G
APOSPC408831-1GEA 2020 EUR
APOSPC408831-1G
1-Nitro-3-(pentafluoroethyl)benzene
1-Nitro-3-(pentafluoroethyl)benzene
Formula: C₈H₄F₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD22053411
CAS Number: 656-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR