4-Fluoro-1-hydroxymethyl-admantane

Supplier: Apollo Scientific

PC430115-1G
APOSPC430115-1GEA 865 EUR
APOSPC430115-1G
4-Fluoro-1-hydroxymethyl-admantane
4-Fluoro-1-hydroxymethyl-admantane
Formula: C₁₁H₁₇FO
MW: 184.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18800685
CAS Number: 1283721-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR