2-[(4-Fluorobenzyl)amino]ethanol

Supplier: Apollo Scientific

PC430520-250MG
APOSPC430520-250MGEA 172 EUR
APOSPC430520-250MG
2-[(4-Fluorobenzyl)amino]ethanol
2-[(4-Fluorobenzyl)amino]ethanol
Formula: C₉H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD04560903
CAS Number: 22116-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR