Methyl-3-(trifluoromethyl)phenylacetate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2-(3-(trifluoromethyl)phenyl)acetate

PC4391-1G PC4391-25G PC4391-5G
APOSPC4391-1GEA 21.9 EUR
APOSPC4391-1G APOSPC4391-25G APOSPC4391-5G
Methyl-3-(trifluoromethyl)phenylacetate
Methyl-3-(trifluoromethyl)phenylacetate
Formula: C₁₀H₉F₃O₂
Boiling Pt: 211 °C
Density: 1.24 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04039762
CAS Number: 62451-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR