sec-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Supplier: Apollo Scientific

PC450476-25G
APOSPC450476-25GEA 1540 EUR
APOSPC450476-25G
sec-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate
sec-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formula: C₉H₁₂F₆O₃
Storage Temperature: Ambient
MDL Number: MFCD28334448
CAS Number: 1980035-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR