4-Chloro-5-fluoro-2-nitroaniline 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-5-chloro-4-fluoronitrobenzene

PC49021-1G
APOSPC49021-1GEA 30.6 EUR
APOSPC49021-1G
4-Chloro-5-fluoro-2-nitroaniline 97%
4-Chloro-5-fluoro-2-nitroaniline
Formula: C₆H₄ClFN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12828238
CAS Number: 428871-64-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR