3-(Pentafluorothio)benzyl bromide 97%

Supplier: Apollo Scientific
Danger

PC50105-1G PC50105-5G
APOSPC50105-1GEA 516 EUR
APOSPC50105-1G APOSPC50105-5G
3-(Pentafluorothio)benzyl bromide 97%
3-(Pentafluorothio)benzyl bromide
Formula: C₇H₆BrF₅S
Storage Temperature: Ambient
MDL Number: MFCD16652411
CAS Number: 1056893-24-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR