3-Fluoro-2-nitrobenzyl bromide 95%

Supplier: Apollo Scientific
Danger

PC501144-1G PC501144-250MG PC501144-5G
APOSPC501144-1GEA 473 EUR
APOSPC501144-1G APOSPC501144-250MG APOSPC501144-5G
3-Fluoro-2-nitrobenzyl bromide 95%
3-Fluoro-2-nitrobenzyl bromide
Formula: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185622
CAS Number: 1104543-73-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR