2,3-Difluoro-6-methoxytoluene 98%

Supplier: Apollo Scientific

Synonyms: 2-Methyl-3,4-difluoroanisole

PC50424-10G PC50424-1G PC50424-5G PC50424-25G PC50424-100G
APOSPC50424-10GEA 574 EUR
APOSPC50424-10G APOSPC50424-1G APOSPC50424-5G APOSPC50424-25G APOSPC50424-100G
2,3-Difluoro-6-methoxytoluene 98%
2,3-Difluoro-6-methoxytoluene
Formula: C₈H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD12922652
CAS Number: 1208078-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR