4-Fluoro-2-(hydroxymethyl)benzimidazole

Supplier: Apollo Scientific

PC510257-1G
APOSPC510257-1GEA 713 EUR
APOSPC510257-1G
4-Fluoro-2-(hydroxymethyl)benzimidazole
4-Fluoro-2-(hydroxymethyl)benzimidazole
Formula: C₈H₇FN₂O
MW: 166.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18807830
CAS Number: 724788-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR