(2-Fluoro-6-nitrophenyl)methanol

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoro-6-nitrobenzyl alcohol

PC51079-1G PC51079-5G
APOSPC51079-1GEA 325 EUR
APOSPC51079-1G APOSPC51079-5G
(2-Fluoro-6-nitrophenyl)methanol
(2-Fluoro-6-nitrophenyl)methanol
Formula: C₇H₆FNO₃
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD11110169
CAS Number: 1643-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR