Methyl heptafluorobutyrate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2,2,3,3,4,4,4-heptafluorobutanoate, Methyl perfluorobutyrate

PC5110-50G PC5110-5G
APOSPC5110-50GEA 154 EUR
APOSPC5110-50G APOSPC5110-5G
Methyl heptafluorobutyrate 98%
Methyl heptafluorobutyrate
Formula: C₅H₃F₇O₂
Boiling Pt: 80…81 °C
Melting Pt: –86 °C
Density: 1.472 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000433
CAS Number: 356-24-1
EINECS: 206-600-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR