4-Bromo-2-fluoro-6-nitroaniline

Supplier: Apollo Scientific
Warning

PC5180-1G PC5180-25G PC5180-5G
APOSPC5180-1GEA 21.9 EUR
APOSPC5180-1G APOSPC5180-25G APOSPC5180-5G
4-Bromo-2-fluoro-6-nitroaniline
4-Bromo-2-fluoro-6-nitroaniline
Formula: C₆H₄BrFN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD03094271
CAS Number: 517920-70-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR