1-Bromo-2-chloro-4-(trifluoromethoxy)benzene

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-4-(Trifluoromethoxy)bromobenzene

PC53328-1G PC53328-25G PC53328-5G PC53328-100G
APOSPC53328-1GEA 20.5 EUR
APOSPC53328-1G APOSPC53328-25G APOSPC53328-5G APOSPC53328-100G
1-Bromo-2-chloro-4-(trifluoromethoxy)benzene
1-Bromo-2-chloro-4-(trifluoromethoxy)benzene
Formula: C₇H₃BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD11111892
CAS Number: 892845-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR