3-Amino-2-chloro-6-(trifluoromethyl)pyridine 98%

Supplier: Apollo Scientific
Warning

PC5588-1G PC5588-5G PC5588-25G
APOSPC5588-1GEA 21.9 EUR
APOSPC5588-1G APOSPC5588-5G APOSPC5588-25G
3-Amino-2-chloro-6-(trifluoromethyl)pyridine 98%
3-Amino-2-chloro-6-(trifluoromethyl)pyridine
Formula: C6H4ClF3N2
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD01862661
CAS Number: 117519-09-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR