2'-Bromo-6'-fluoro-3'-(trifluoromethyl)acetophenone 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1-[2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl]ethanone

PC56259-1G PC56259-250MG PC56259-5G
APOSPC56259-1GEA 696 EUR
APOSPC56259-1G APOSPC56259-250MG APOSPC56259-5G
2'-Bromo-6'-fluoro-3'-(trifluoromethyl)acetophenone 95%
2'-Bromo-6'-fluoro-3'-(trifluoromethyl)acetophenone
Formula: C₉H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD31613606
CAS Number: 2091610-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR