2-Bromobenzyl fluoride 95%

Supplier: Apollo Scientific
Danger

PC56790-1G PC56790-250MG
APOSPC56790-1GEA 609 EUR
APOSPC56790-1G APOSPC56790-250MG
2-Bromobenzyl fluoride 95%
2-Bromobenzyl fluoride
Formula: C₇H₆BrF
Storage Temperature: Ambient
MDL Number: MFCD26726839
CAS Number: 446-47-9
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR