3-Chloro-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}aniline

Supplier: Apollo Scientific

PC6052-1G
APOSPC6052-1GEA 314 EUR
APOSPC6052-1G
3-Chloro-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}aniline
3-Chloro-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}aniline
Formula: C₁₂H₈ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD11109516
CAS Number: 1211669-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR