Tetrabutylammonium tetrafluoroborate 97%

Supplier: Apollo Scientific
Warning

Synonyms: Tetrabutylammoniumtetrafluoroborate

PC6635-100G PC6635-25G PC6635-1G
APOSPC6635-100GEA 106 EUR
APOSPC6635-100G APOSPC6635-25G APOSPC6635-1G
Tetrabutylammonium tetrafluoroborate 97%
Tetrabutylammonium tetrafluoroborate

Electrolyte additive used in the synthesis of electrically conducting poly(thiophene) polymers, JOC., 1994,59, 442

Formula: C₁₆H₃₆BF₄N
Melting Pt: 155…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS Number: 429-42-5
EINECS: 207-058-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR